Product Details

SNP ID
rs2177464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99557778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTTCTTGTGTTTAATTTGAAAC[A/G]TCCAGTAACATTAAAGAACAGCATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM200A PubMed Links

Gene Details

Gene
FAM200A
Gene Name
family with sequence similarity 200 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145111.3 44 Intron NP_659802.1
Gene
ZNF655
Gene Name
zinc finger protein 655
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009958.1 44 Intron NP_001009958.1
NM_001009960.1 44 Intron NP_001009960.1
NM_001083956.1 44 Intron NP_001077425.1
NM_001085366.1 44 Intron NP_001078835.1
NM_001085367.1 44 Intron NP_001078836.1
NM_001085368.1 44 Intron NP_001078837.1
NM_024061.3 44 Intron NP_076966.1
NM_138494.2 44 Intron NP_612503.1
XM_017012598.1 44 UTR 5 XP_016868087.1
XM_017012599.1 44 Intron XP_016868088.1
XM_017012600.1 44 Intron XP_016868089.1
XM_017012601.1 44 Intron XP_016868090.1
XM_017012602.1 44 UTR 5 XP_016868091.1
XM_017012603.1 44 UTR 5 XP_016868092.1
XM_017012604.1 44 Intron XP_016868093.1
XM_017012605.1 44 Intron XP_016868094.1
XM_017012606.1 44 Intron XP_016868095.1
XM_017012607.1 44 Intron XP_016868096.1
XM_017012608.1 44 UTR 5 XP_016868097.1
XM_017012609.1 44 Intron XP_016868098.1
XM_017012610.1 44 Intron XP_016868099.1
XM_017012611.1 44 UTR 5 XP_016868100.1
XM_017012612.1 44 UTR 5 XP_016868101.1
XM_017012613.1 44 Intron XP_016868102.1

View Full Product Details