Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001031703.2 | Intron | NP_001026873.2 | |||
XM_005265241.3 | Intron | XP_005265298.1 | |||
XM_011533851.2 | Intron | XP_011532153.1 | |||
XM_011533852.2 | Intron | XP_011532154.1 | |||
XM_011533853.2 | Intron | XP_011532155.1 | |||
XM_011533854.2 | Intron | XP_011532156.1 | |||
XM_011533856.2 | Intron | XP_011532158.1 | |||
XM_011533857.2 | Intron | XP_011532159.1 | |||
XM_017006669.1 | Intron | XP_016862158.1 | |||
XM_017006670.1 | Intron | XP_016862159.1 |