Product Details

SNP ID
rs2896336
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123818398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCTAGACTGTCTTGTGAATTC[A/G]TACTCATTCTCAAGAGTCTACTGAT
Phenotype
MIM: 604510
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HYAL4 PubMed Links
Additional Information
For this assay, SNP(s) [rs116278459] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HYAL4
Gene Name
hyaluronoglucosaminidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012269.2 Intron NP_036401.2
XM_011515990.2 Intron XP_011514292.1
XM_011515991.2 Intron XP_011514293.1
XM_017011911.1 Intron XP_016867400.1
Gene
HYALP1
Gene Name
hyaluronoglucosaminidase pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details