Product Details

SNP ID
rs2074105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44315880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAGAGTGGCCATCCAACCTTACT[C/G]CAGGGACCCCACGTCACTCACAAGG
Phenotype
MIM: 605448 MIM: 610820
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RUNDC3A PubMed Links

Gene Details

Gene
RUNDC3A
Gene Name
RUN domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144825.1 Intron NP_001138297.1
NM_001144826.1 Intron NP_001138298.1
NM_006695.4 Intron NP_006686.1
XM_005256964.3 Intron XP_005257021.1
XM_017024035.1 Intron XP_016879524.1
XM_017024036.1 Intron XP_016879525.1
XM_017024037.1 Intron XP_016879526.1
XM_017024038.1 Intron XP_016879527.1
XM_017024039.1 Intron XP_016879528.1
XM_017024040.1 Intron XP_016879529.1
Gene
RUNDC3A-AS1
Gene Name
RUNDC3A antisense RNA 1
There are no transcripts associated with this gene.

Gene
SLC25A39
Gene Name
solute carrier family 25 member 39
There are no transcripts associated with this gene.

View Full Product Details