Product Details
- SNP ID
-
rs2220091
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:144684400 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGACTCCAAGATCTCCTTTGCTT[A/C]TTTTTTTCTAGACATGTTTGAGGCC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TEX41
PubMed Links
Gene Details
- Gene
- TEX41
- Gene Name
- testis expressed 41 (non-protein coding)
There are no transcripts associated with this gene.
View Full Product Details