Product Details

SNP ID
rs2220576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:44179220 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCGAAAAAGTCATCTATATCCCT[C/G]TCTTCTCCTCCCTCTTGTCTTTCCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCTD8 PubMed Links

Gene Details

Gene
KCTD8
Gene Name
potassium channel tetramerization domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198353.2 Intron NP_938167.1
XM_011513690.2 Intron XP_011511992.1

View Full Product Details