Product Details
- SNP ID
-
rs2221165
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:117151451 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTCTAATTCACTCTTCAACTCTC[C/T]GTGCAGAGCTAGTAATTAGAAAAGC
- Phenotype
-
MIM: 609095
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74829496,rs75987440] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXO21
- Gene Name
- F-box protein 21
- Gene
- TESC-AS1
- Gene Name
- TESC antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details