Product Details

SNP ID
rs2913399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:122064971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTTTGTTTTTTAAGAAAGCAA[C/G]TATATTAAAATATCTCTGTTAGAAA
Phenotype
MIM: 153455 MIM: 610479
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOX PubMed Links

Gene Details

Gene
LOX
Gene Name
lysyl oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178102.2 2369 UTR 3 NP_001171573.1
NM_001317073.1 2369 UTR 3 NP_001304002.1
NM_002317.6 2369 UTR 3 NP_002308.2
Gene
SRFBP1
Gene Name
serum response factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152546.2 2369 Intron NP_689759.2
XM_017009111.1 2369 Intron XP_016864600.1

View Full Product Details