Product Details

SNP ID
rs2238007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117213764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGTGGCAGTGCAAATGAGTACA[C/T]TAGAACTCTCTCTACCAGAACTAAA
Phenotype
MIM: 604872 MIM: 600818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCSK7 PubMed Links

Gene Details

Gene
PCSK7
Gene Name
proprotein convertase subtilisin/kexin type 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004716.3 Intron NP_004707.2
XM_006718940.3 Intron XP_006719003.1
XM_017018546.1 Intron XP_016874035.1
Gene
TAGLN
Gene Name
transgelin
There are no transcripts associated with this gene.

View Full Product Details