Product Details

SNP ID
rs2239968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:25005946 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCCCGCGGGGCCAATTCCGGAT[A/T]TCCAGGGTTAGAAATGAGCCCACCT
Phenotype
MIM: 300382 MIM: 312040
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARX PubMed Links

Gene Details

Gene
ARX
Gene Name
aristaless related homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139058.2 Intron NP_620689.1
Gene
POLA1
Gene Name
DNA polymerase alpha 1, catalytic subunit
There are no transcripts associated with this gene.

View Full Product Details