Product Details

SNP ID
rs2266951
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:20919153 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACCTTTAACCAAAATATCAGAACT[A/G]TATATTGATACTGTGGTGCCAAGTT
Phenotype
MIM: 602007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRKL PubMed Links

Gene Details

Gene
CRKL
Gene Name
CRK like proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005207.3 Intron NP_005198.1

View Full Product Details