Product Details
- SNP ID
-
rs2286005
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3726246 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTCTGGGAGTCGTGTACGGGGGG[G/T]GGGAGCCGCCTGAACACGGGAAGAA
- Phenotype
-
MIM: 600140
MIM: 606219
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CREBBP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149315725] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CREBBP
- Gene Name
- CREB binding protein
- Gene
- TRAP1
- Gene Name
- TNF receptor associated protein 1
There are no transcripts associated with this gene.
View Full Product Details