Product Details

SNP ID
rs2964975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99141840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCGGTTACTCTCAGAGCTGCTTT[C/G]GGGCGCAGCTCCTGCTGCAGCCAGG
Phenotype
MIM: 607166
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C2orf15 PubMed Links

Gene Details

Gene
C2orf15
Gene Name
chromosome 2 open reading frame 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317992.1 228 UTR 5 NP_001304921.1
NM_144706.3 228 UTR 5 NP_653307.1
Gene
TSGA10
Gene Name
testis specific 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025244.2 228 Intron NP_079520.1
NM_182911.3 228 Intron NP_878915.2
XM_005264023.1 228 Intron XP_005264080.1
XM_005264024.2 228 Intron XP_005264081.1
XM_005264025.1 228 Intron XP_005264082.1
XM_005264026.1 228 Intron XP_005264083.1
XM_005264027.1 228 Intron XP_005264084.1
XM_005264029.1 228 Intron XP_005264086.1
XM_006712781.2 228 Intron XP_006712844.1
XM_011511934.1 228 Intron XP_011510236.1
XM_011511935.2 228 Intron XP_011510237.1
XM_011511937.1 228 Intron XP_011510239.1
XM_011511939.1 228 Intron XP_011510241.1
XM_011511940.2 228 Intron XP_011510242.1
XM_011511941.1 228 Intron XP_011510243.1
XM_011511942.1 228 Intron XP_011510244.1
XM_011511943.2 228 Intron XP_011510245.1
XM_017005035.1 228 Intron XP_016860524.1
XM_017005036.1 228 Intron XP_016860525.1
XM_017005037.1 228 Intron XP_016860526.1
XM_017005038.1 228 Intron XP_016860527.1
XM_017005039.1 228 Intron XP_016860528.1
XM_017005040.1 228 Intron XP_016860529.1
XM_017005041.1 228 Intron XP_016860530.1
XM_017005042.1 228 Intron XP_016860531.1
XM_017005043.1 228 Intron XP_016860532.1
XM_017005044.1 228 Intron XP_016860533.1
XM_017005045.1 228 Intron XP_016860534.1
XM_017005046.1 228 Intron XP_016860535.1
XM_017005047.1 228 Intron XP_016860536.1
XM_017005048.1 228 Intron XP_016860537.1

View Full Product Details