Product Details

SNP ID
rs2286611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64236185 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGGCTGTTGGGTGAGCTTTTCTC[C/T]CTTCAGTCTTAGAGCATCCCCTTTC
Phenotype
MIM: 604189 MIM: 186946 MIM: 601140 MIM: 610470 MIM: 601398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
There are no transcripts associated with this gene.

Gene
FKBP2
Gene Name
FK506 binding protein 2
There are no transcripts associated with this gene.

Gene
LOC105369340
Gene Name
uncharacterized LOC105369340
There are no transcripts associated with this gene.

Gene
NUDT22
Gene Name
nudix hydrolase 22
There are no transcripts associated with this gene.

Gene
PPP1R14B
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14B
There are no transcripts associated with this gene.

Gene
TRPT1
Gene Name
tRNA phosphotransferase 1
There are no transcripts associated with this gene.

Gene
VEGFB
Gene Name
vascular endothelial growth factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243733.1 Intron NP_001230662.1
NM_003377.4 Intron NP_003368.1

View Full Product Details