Product Details

SNP ID
rs2273905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102508662 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACACGTTAGCTCCTGGGAGGAGA[C/T]AGGAGGGTGAAAACAACCTGGAGAC
Phenotype
MIM: 615000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD9 PubMed Links

Gene Details

Gene
ANKRD9
Gene Name
ankyrin repeat domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152326.2 585 UTR 5 NP_689539.1
XM_005267310.2 585 UTR 5 XP_005267367.1
XM_005267311.3 585 Intron XP_005267368.1
Gene
TECPR2
Gene Name
tectonin beta-propeller repeat containing 2
There are no transcripts associated with this gene.

View Full Product Details