Product Details

SNP ID
rs2274664
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:54238956 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CCTTCCTCTCCCACCCCCTGCCGCC[C/T]ATGAAATGGGAACTGAGTTGCCCAA
Phenotype
MIM: 607390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSBP3 PubMed Links

Gene Details

Gene
SSBP3
Gene Name
single stranded DNA binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009955.3 Intron NP_001009955.1
NM_018070.4 Intron NP_060540.2
NM_145716.3 Intron NP_663768.1
XM_006710545.3 Intron XP_006710608.1
XM_017000897.1 Intron XP_016856386.1
XM_017000898.1 Intron XP_016856387.1
XM_017000899.1 Intron XP_016856388.1
Gene
SSBP3-AS1
Gene Name
SSBP3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details