Product Details

SNP ID
rs2293585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89822445 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGAAACCCTGGAGCACCCCGGGC[C/T]GGAACCCTTCCCTCAGATCTGTCGT
Phenotype
MIM: 607139 MIM: 609217
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FANCA PubMed Links

Gene Details

Gene
FANCA
Gene Name
Fanconi anemia complementation group A
There are no transcripts associated with this gene.

Gene
LOC107984817
Gene Name
uncharacterized LOC107984817
There are no transcripts associated with this gene.

Gene
SPIRE2
Gene Name
spire type actin nucleation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032451.1 Intron NP_115827.1
XM_011523379.1 Intron XP_011521681.1
XM_011523380.2 Intron XP_011521682.1
XM_011523381.1 Intron XP_011521683.1
XM_011523384.1 Intron XP_011521686.1
XM_011523385.1 Intron XP_011521687.1
XM_017023779.1 Intron XP_016879268.1
XM_017023780.1 Intron XP_016879269.1
XM_017023781.1 Intron XP_016879270.1

View Full Product Details