Product Details

SNP ID
rs2298705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95769146 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATTCACAGGCTATTAATACATAA[A/G]TAACTTGAGTCAAAATGACCCTTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM76B PubMed Links

Gene Details

Gene
FAM76B
Gene Name
family with sequence similarity 76 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144664.4 3504 UTR 3 NP_653265.3
XM_005273775.4 3504 UTR 3 XP_005273832.1
XM_005273776.4 3504 UTR 3 XP_005273833.1
XM_011542610.2 3504 UTR 3 XP_011540912.1
XM_011542611.2 3504 UTR 3 XP_011540913.1
XM_011542612.1 3504 UTR 3 XP_011540914.1
XM_011542613.1 3504 UTR 3 XP_011540915.1

View Full Product Details