Product Details

SNP ID
rs2306101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69842433 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGACACAAGTATGAGGAGCAGA[G/T]AGGTTTCTCTTCTTCAGTCTCCACT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00593 PubMed Links

Gene Details

Gene
LINC00593
Gene Name
long intergenic non-protein coding RNA 593
There are no transcripts associated with this gene.

View Full Product Details