Product Details

SNP ID
rs2306589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36493030 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGGAAGGAACAGCTTCCTTAGCC[C/T]TGAGCATCAGGAGTGGGGCCATGTC
Phenotype
MIM: 604500
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO19 PubMed Links

Gene Details

Gene
MYO19
Gene Name
myosin XIX
There are no transcripts associated with this gene.

Gene
ZNHIT3
Gene Name
zinc finger HIT-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281432.1 Intron NP_001268361.1
NM_001281433.1 Intron NP_001268362.1
NM_001281434.1 Intron NP_001268363.1
NM_004773.3 Intron NP_004764.1

View Full Product Details