Product Details

SNP ID
rs3015331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103274801 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACTTCCACAGGTGCTTCTCCCTA[A/C]ATTTAAGGAGGATGTGCTGAGAAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TCEAL5 PubMed Links

Gene Details

Gene
TCEAL5
Gene Name
transcription elongation factor A like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012979.2 Intron NP_001012997.1

View Full Product Details