Product Details
- SNP ID
-
hCV16196210
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:200921114 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATCTGGCTATCCAAATTGTGGAT[A/G]AGAAGGAAGAGTTCTAAAGAAGAAT
- Phenotype
-
MIM: 601182
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ORC2
PubMed Links
Gene Details
- Gene
- ORC2
- Gene Name
- origin recognition complex subunit 2
There are no transcripts associated with this gene.
View Full Product Details