Product Details

SNP ID
rs3025638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29605144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGATCATCCTCAAATATAGATT[A/G]AGAAAAATCTCAAACTGTCCCAAAC
Phenotype
MIM: 603540
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABBR1 PubMed Links

Gene Details

Gene
GABBR1
Gene Name
gamma-aminobutyric acid type B receptor subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319053.1 Intron NP_001305982.1
NM_001470.3 Intron NP_001461.1
NM_021903.2 Intron NP_068703.1
NM_021904.3 Intron NP_068704.2
XM_005248982.2 Intron XP_005249039.1
XM_006715047.3 Intron XP_006715110.1
XM_011514453.2 Intron XP_011512755.1
XM_011514455.1 Intron XP_011512757.1
XM_017010676.1 Intron XP_016866165.1

View Full Product Details