Product Details

SNP ID
rs1805764
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8942805 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAATAATCACCATTAAGTATATG[A/C]TTCCATTTCTCTAGAAGTTCTCATA
Phenotype
MIM: 154540 MIM: 602978
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
M6PR PubMed Links

Gene Details

Gene
M6PR
Gene Name
mannose-6-phosphate receptor, cation dependent
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207024.1 Intron NP_001193953.1
NM_002355.3 Intron NP_002346.1
XM_005253376.2 Intron XP_005253433.1
Gene
PHC1
Gene Name
polyhomeotic homolog 1
There are no transcripts associated with this gene.

View Full Product Details