Product Details
- SNP ID
-
rs2427776
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:51038103 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGAGTTCGAGACCCTGTTTCTAC[A/C]ATAACATATAAAAATTAGGTGTAGT
- Phenotype
-
MIM: 600993
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SMAD4
PubMed Links
Gene Details
- Gene
- SMAD4
- Gene Name
- SMAD family member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005359.5 |
|
Intron |
|
|
NP_005350.1 |
View Full Product Details