Product Details

SNP ID
rs2583088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:69467654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTAATTGATTTGCAGATCTGTTT[A/G]TGAAAGTTTTACAAGTCATGGAAAA
Phenotype
MIM: 610012
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SULF1 PubMed Links

Gene Details

Gene
SULF1
Gene Name
sulfatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128204.1 Intron NP_001121676.1
NM_001128205.1 Intron NP_001121677.1
NM_001128206.1 Intron NP_001121678.1
NM_015170.2 Intron NP_055985.2
XM_006716438.2 Intron XP_006716501.1
XM_006716439.2 Intron XP_006716502.1
XM_006716440.3 Intron XP_006716503.1
XM_006716441.1 Intron XP_006716504.1
XM_006716442.1 Intron XP_006716505.1
XM_011517494.1 Intron XP_011515796.1
XM_011517495.1 Intron XP_011515797.1
XM_017013250.1 Intron XP_016868739.1

View Full Product Details