Product Details

SNP ID
rs2707634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99245387 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCACGCAATGTCTCTGCCCGACTT[C/T]TTTGATCAGAGTCTGGTAAGGTTTT
Phenotype
MIM: 616547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYG2 PubMed Links

Gene Details

Gene
LYG2
Gene Name
lysozyme g2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175735.3 369 Missense Mutation AAA,GAA K86E NP_783862.2
XM_017003751.1 369 Missense Mutation AAA,GAA K86E XP_016859240.1

View Full Product Details