Product Details
- SNP ID
-
rs2697655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:10760011 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAGGACTTGGGTACAGGGATCTGT[A/G]ATAACATTAACTGAGGAAAATAGGG
- Phenotype
-
MIM: 600280
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NUBP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115914510] are located under a probe and SNP(s) [rs74988634] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NUBP1
- Gene Name
- nucleotide binding protein 1
- Gene
- TVP23A
- Gene Name
- trans-golgi network vesicle protein 23 homolog A
View Full Product Details