Product Details

SNP ID
rs2742689
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:3078698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGCAGGACACTGCCAACGTTT[G/T]AGCGGAGGGAGGAAATGACGGGGTG
Phenotype
MIM: 605557
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRDM16 PubMed Links

Gene Details

Gene
PRDM16
Gene Name
PR domain 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022114.3 Intron NP_071397.3
NM_199454.2 Intron NP_955533.2
XM_005244772.4 Intron XP_005244829.1
XM_005244773.4 Intron XP_005244830.1
XM_005244774.4 Intron XP_005244831.1
XM_006710814.3 Intron XP_006710877.1
XM_011541945.2 Intron XP_011540247.1
XM_017002050.1 Intron XP_016857539.1

View Full Product Details