Product Details
- SNP ID
-
rs2753602
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:39175783 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACGCAACAAGCCGCCTTACCCCA[A/G]CTTCCTGTCCACGTGCAGCCTCCTC
- Phenotype
-
MIM: 603154
MIM: 610397
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PNN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17108939] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PNN
- Gene Name
- pinin, desmosome associated protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002687.3 |
|
Intron |
|
|
NP_002678.2 |
- Gene
- TRAPPC6B
- Gene Name
- trafficking protein particle complex 6B
There are no transcripts associated with this gene.
View Full Product Details