Product Details

SNP ID
rs2149269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:107169568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCTAGATAGCTGCTCCTGTCCA[A/G]CTGGACCAGCAAGAAAGGGCTGGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM155A PubMed Links

Gene Details

Gene
FAM155A
Gene Name
family with sequence similarity 155 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080396.2 2822 UTR 3 NP_001073865.1
XM_011521109.2 2822 Intron XP_011519411.1

View Full Product Details