Product Details

SNP ID
rs2586238
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:999082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTGAAAGTTGTTACTAGAAAGCA[A/G]ATCAGAAGCATCAGATACATTCTAG
Phenotype
MIM: 600365 MIM: 607251
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ABR PubMed Links

Gene Details

Gene
ABR
Gene Name
active BCR-related
There are no transcripts associated with this gene.

Gene
TIMM22
Gene Name
translocase of inner mitochondrial membrane 22 homolog (yeast)
There are no transcripts associated with this gene.

View Full Product Details