Product Details

SNP ID
rs41281234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3047800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGGTGGAAGCAGCATTGGACTC[C/T]CGGAAGAACTACAACTGGGCCATCA
Phenotype
MIM: 602352 MIM: 611988 MIM: 176884
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNRH2 PubMed Links

Gene Details

Gene
GNRH2
Gene Name
gonadotropin releasing hormone 2
There are no transcripts associated with this gene.

Gene
MRPS26
Gene Name
mitochondrial ribosomal protein S26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030811.3 589 Silent Mutation TCC,TCT S183S NP_110438.1
Gene
PTPRA
Gene Name
protein tyrosine phosphatase, receptor type A
There are no transcripts associated with this gene.

View Full Product Details