Product Details
- SNP ID
-
rs3732839
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:138162423 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGCCAAGTTGGGCACAAAATTCAGT[A/T]GTCTGAGGATTGATCCTATGAATCA
- Phenotype
-
MIM: 607024
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DBR1
PubMed Links
Gene Details
- Gene
- DBR1
- Gene Name
- debranching RNA lariats 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016216.3 |
1255 |
Silent Mutation |
ACA,ACT |
T367T |
NP_057300.2 |
View Full Product Details