Product Details
- SNP ID
-
rs2544809
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:177458498 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAATGGTGTCAGCTGCATCGTGGA[C/T]CTCCGTGGCGTCAGCTGTGGCAGGC
- Phenotype
-
MIM: 126660
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DBN1
PubMed Links
Gene Details
- Gene
- DBN1
- Gene Name
- drebrin 1
- Gene
- PRR7
- Gene Name
- proline rich 7 (synaptic)
There are no transcripts associated with this gene.
View Full Product Details