Product Details

SNP ID
rs2246945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138124634 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGGGCCTTGGTTGCCCCAAATG[G/T]CTGAATTATAGTGTTCAACAAAGTT
Phenotype
MIM: 616709
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
A4GNT PubMed Links

Gene Details

Gene
A4GNT
Gene Name
alpha-1,4-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016161.2 855 Missense Mutation GAC,GCC D218A NP_057245.1
XM_017006543.1 855 Missense Mutation GAC,GCC D218A XP_016862032.1
XM_017006544.1 855 Missense Mutation GAC,GCC D218A XP_016862033.1
Gene
DZIP1L
Gene Name
DAZ interacting zinc finger protein 1 like
There are no transcripts associated with this gene.

View Full Product Details