Product Details

SNP ID
rs3788441
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:31937797 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACTCTCTGAAGGTCAGGGGCAGTG[C/G]TGGGAAGAATGGTGAAGATGGAGCA
Phenotype
MIM: 113508
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C22orf24 PubMed Links

Gene Details

Gene
C22orf24
Gene Name
chromosome 22 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302819.1 Intron NP_001289748.1
NM_001302820.1 Intron NP_001289749.1
NM_015372.2 Intron NP_056187.1
Gene
YWHAH
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta
There are no transcripts associated with this gene.

View Full Product Details