Product Details
- SNP ID
-
rs6060263
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31605307 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTCTCATTCCACGTTCTTAACTG[C/T]TCCATTTTCCGTATCTGCTTCGGGC
- Phenotype
-
MIM: 600349
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ID1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111423898] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ID1
- Gene Name
- inhibitor of DNA binding 1, HLH protein
- Gene
- MIR3193
- Gene Name
- microRNA 3193
There are no transcripts associated with this gene.
View Full Product Details