Product Details

SNP ID
rs12564525
Assay Type
DME
NCBI dbSNP Submissions
44
Location
Chr.1:47141609 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
TGGATCCTACAAATTCCTGGCTCCA[C/T]GGATTGGTATGTGTGCAAACTAGGA
Phenotype
MIM: 615341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
CYP4A22*11,c.376C>T CYP4A22*11,g.4124C>T CYP4A22*12A,c.376C>T CYP4A22*12A,g.4124C>T CYP4A22*12B,c.376C>T CYP4A22*12B,g.4124C>T CYP4A22*13A,c.376C>T CYP4A22*13A,g.4124C>T CYP4A22*13B,c.376C>T CYP4A22*13B,g.4124C>T CYP4A22*14,c.376C>T CYP4A22*14,g.4124C>T CYP4A22*15,c.376C>T CYP4A22*15,g.4124C>T CYP4A22*8,c.376C>T CYP4A22*8,g.4124C>T CYP4A22*9,c.376C>T CYP4A22*9,g.4124C>T
Literature Links
CYP4A22 PubMed Links

Gene Details

Gene
CYP4A22
Gene Name
cytochrome P450 family 4 subfamily A member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010969.3 Intron NP_001010969.2
NM_001308102.1 Intron NP_001295031.1
XM_005270767.3 Intron XP_005270824.1
XM_005270770.3 Intron XP_005270827.1

View Full Product Details