Product Details

SNP ID
hCV25472936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53425437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGCATTTGCACCCTGACCCTAA[A/G]GCTCTTGTCTGTTCCAGCCCTGCTA
Phenotype
MIM: 600956 MIM: 189906
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMHR2 PubMed Links

Gene Details

Gene
AMHR2
Gene Name
anti-Mullerian hormone receptor type 2
There are no transcripts associated with this gene.

Gene
SP1
Gene Name
Sp1 transcription factor
There are no transcripts associated with this gene.

View Full Product Details