Product Details

SNP ID
rs7563771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86785717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCACAACAGTATTGTGACCCTT[A/G]TGGTGTACTGTAGATTTTACCTAGT
Phenotype
MIM: 186910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD8A PubMed Links

Gene Details

Gene
CD8A
Gene Name
CD8a molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145873.1 1942 UTR 3 NP_001139345.1
NM_001768.6 1942 UTR 3 NP_001759.3
NM_171827.3 1942 UTR 3 NP_741969.1
Gene
RMND5A
Gene Name
required for meiotic nuclear division 5 homolog A
There are no transcripts associated with this gene.

View Full Product Details