Product Details

SNP ID
rs11654834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40632096 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAATCAGGAACAGGTTAGATTGA[C/T]ATATTTAACAGGTAGATTTAGGCAT
Phenotype
MIM: 603111
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMARCE1 PubMed Links

Gene Details

Gene
SMARCE1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003079.4 Intron NP_003070.3

View Full Product Details