Product Details

SNP ID
rs9904341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78214286 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTAACCGCCAGATTTGAATCGC[C/G]GGACCCGTTGGCAGAGGTGGCGGCG
Phenotype
MIM: 603352
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AFMID PubMed Links

Gene Details

Gene
AFMID
Gene Name
arylformamidase
There are no transcripts associated with this gene.

Gene
BIRC5
Gene Name
baculoviral IAP repeat containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012270.1 91 UTR 5 NP_001012270.1
NM_001012271.1 91 UTR 5 NP_001012271.1
NM_001168.2 91 UTR 5 NP_001159.2

View Full Product Details