Product Details

SNP ID
rs60938534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73136571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTATGATCCTTTATCCTACCAGA[C/G]TTAGGAAAACAATTAACATTCTTAT
Phenotype
MIM: 616464 MIM: 609233
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ECD PubMed Links

Gene Details

Gene
ECD
Gene Name
ecdysoneless cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135752.1 Intron NP_001129224.1
NM_001135753.1 Intron NP_001129225.1
NM_007265.2 Intron NP_009196.1
Gene
NUDT13
Gene Name
nudix hydrolase 13
There are no transcripts associated with this gene.

View Full Product Details