Product Details

SNP ID
rs7356212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:166735268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTATCACATGTTAAAAATCTA[A/T]CATTTGTTAAAATGTATGTGCTAAA
Phenotype
MIM: 607989
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SPOCK3 PubMed Links

Gene Details

Gene
SPOCK3
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040159.1 Intron NP_001035249.1
NM_001204352.1 Intron NP_001191281.1
NM_001204353.1 Intron NP_001191282.1
NM_001204354.1 Intron NP_001191283.1
NM_001204355.1 Intron NP_001191284.1
NM_001204356.1 Intron NP_001191285.1
NM_001204358.1 Intron NP_001191287.1
NM_001251967.1 Intron NP_001238896.1
NM_016950.2 Intron NP_058646.2
XM_011532018.1 Intron XP_011530320.1
XM_017008257.1 Intron XP_016863746.1
XM_017008258.1 Intron XP_016863747.1

View Full Product Details