Product Details

SNP ID
hCV25597401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52899701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATAAAATTGTTGAATAGAGAATT[C/T]TATACTGGGGTCTAGTAAATTGAAA
Phenotype
MIM: 605449
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTA3 PubMed Links

Gene Details

Gene
GSTA3
Gene Name
glutathione S-transferase alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000847.4 Intron NP_000838.3

View Full Product Details