Product Details

SNP ID
rs62620010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39175214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCCCGGCTGTCAGTCCTTTCGC[A/G]CCTCGGCGGCGCGGCATAGCCCGGC
Phenotype
MIM: 603154 MIM: 610397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNN PubMed Links

Gene Details

Gene
PNN
Gene Name
pinin, desmosome associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002687.3 32 UTR 5 NP_002678.2
Gene
TRAPPC6B
Gene Name
trafficking protein particle complex 6B
There are no transcripts associated with this gene.

View Full Product Details