Product Details

SNP ID
rs79121594
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112644584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTGAGTCCATTATTCTCTCAG[A/G]CAGTTAAAGGCTAATCTAAATATTT
Phenotype
MIM: 612026 MIM: 614596 MIM: 614597 MIM: 614598 MIM: 614599 MIM: 614600
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LARP7 PubMed Links

Gene Details

Gene
LARP7
Gene Name
La ribonucleoprotein domain family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267039.1 Intron NP_001253968.1
NM_015454.2 Intron NP_056269.1
NM_016648.3 Intron NP_057732.2
Gene
MIR302A
Gene Name
microRNA 302a
There are no transcripts associated with this gene.

Gene
MIR302B
Gene Name
microRNA 302b
There are no transcripts associated with this gene.

Gene
MIR302C
Gene Name
microRNA 302c
There are no transcripts associated with this gene.

Gene
MIR302D
Gene Name
microRNA 302d
There are no transcripts associated with this gene.

Gene
MIR367
Gene Name
microRNA 367
There are no transcripts associated with this gene.

Gene
ZGRF1
Gene Name
zinc finger GRF-type containing 1
There are no transcripts associated with this gene.

View Full Product Details