Product Details

SNP ID
rs28440970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64227440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATTTTAACTCTTTCTTTAAAAT[C/T]ATTTTTTTCATGGATTACAATGATC
Phenotype
MIM: 601663 MIM: 608442
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESR2 PubMed Links

Gene Details

Gene
ESR2
Gene Name
estrogen receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040275.1 3309 UTR 3 NP_001035365.1
NM_001214902.1 3309 UTR 3 NP_001201831.1
NM_001271876.1 3309 Intron NP_001258805.1
NM_001271877.1 3309 Intron NP_001258806.1
NM_001291712.1 3309 UTR 3 NP_001278641.1
NM_001291723.1 3309 UTR 3 NP_001278652.1
NM_001437.2 3309 Intron NP_001428.1
XM_017021079.1 3309 Intron XP_016876568.1
XM_017021080.1 3309 Intron XP_016876569.1
XM_017021081.1 3309 Intron XP_016876570.1
XM_017021082.1 3309 Intron XP_016876571.1
XM_017021083.1 3309 Intron XP_016876572.1
XM_017021084.1 3309 Intron XP_016876573.1
Gene
SYNE2
Gene Name
spectrin repeat containing nuclear envelope protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015180.4 3309 Intron NP_055995.4
NM_182910.2 3309 Intron NP_878914.1
NM_182913.2 3309 Intron NP_878917.1
NM_182914.2 3309 Intron NP_878918.2
XM_005267454.1 3309 Intron XP_005267511.1
XM_005267456.1 3309 Intron XP_005267513.1
XM_005267457.1 3309 Intron XP_005267514.1
XM_005267458.1 3309 Intron XP_005267515.1
XM_005267459.1 3309 Intron XP_005267516.1
XM_011536574.1 3309 Intron XP_011534876.1
XM_011536575.2 3309 Intron XP_011534877.1
XM_011536576.2 3309 Intron XP_011534878.1
XM_011536577.2 3309 Intron XP_011534879.1
XM_011536578.1 3309 Intron XP_011534880.1
XM_011536579.1 3309 Intron XP_011534881.1
XM_011536580.1 3309 Intron XP_011534882.1
XM_011536581.1 3309 Intron XP_011534883.1
XM_011536582.1 3309 Intron XP_011534884.1
XM_011536584.2 3309 Intron XP_011534886.1
XM_017021101.1 3309 Intron XP_016876590.1
XM_017021102.1 3309 Intron XP_016876591.1
XM_017021103.1 3309 Intron XP_016876592.1
XM_017021104.1 3309 Intron XP_016876593.1

View Full Product Details