Product Details

SNP ID
rs148584514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65920214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGAGGGAGCCACCTGAGTAAA[C/G]CAGGGCAGGCCCGGGAGCGGAGATC
Phenotype
MIM: 602289
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C11orf68 PubMed Links

Gene Details

Gene
C11orf68
Gene Name
chromosome 11 open reading frame 68
There are no transcripts associated with this gene.

Gene
DRAP1
Gene Name
DR1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006442.3 Intron NP_006433.2

View Full Product Details